4-Butoxy-2,5-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37854-500MG 37854-1G 37854-5G
77338-490EA 491.31 USD
77338-490 77350-178 77374-382
4-Butoxy-2,5-difluorobenzoic acid ≥97%
4-Butoxy-2,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₃
MW: 230.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403743
CAS Number: 2624417-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR