3-Methoxy-4-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

37762-500MG 37762-1G 37762-5G 37762-25G
77326-502EA 107.12 USD
77326-502 77343-146 77362-800 77374-378
3-Methoxy-4-methylbenzamide ≥97%
3-Methoxy-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797809
CAS Number: 1017082-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR