3-Methoxy-N,4-dimethylbenzamide ≥97%
Supplier: AOBChem USA
|
Warning
|
37761-500MG
37761-1G
37761-5G
37761-25G
77326-500EA
198.79
USD
77326-500
77350-176
77362-798
77385-622
3-Methoxy-N,4-dimethylbenzamide ≥97%
3-Methoxy-N,4-dimethylbenzamide
Precaución: for Research Use Only
Formula:
C₁₀H₁₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD11767633 CAS Number: 1090859-40-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...