(3-Methoxy-4-methylphenyl)(thiomorpholino)methanone ≥95%
Supplier: AOBChem USA
|
Warning
|
37757-500MG
37757-1G
37757-5G
37757-25G
77326-496EA
234.84
USD
77326-496
77350-172
77362-792
77385-618
(3-Methoxy-4-methylphenyl)(thiomorpholino)methanone ≥95%
(3-Methoxy-4-methylphenyl)(thiomorpholino)methanone
Precaución: for Research Use Only
Formula:
C₁₃H₁₇NO₂S MW: 251.345 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD33403741 CAS Number: 2635937-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...