(3-Methoxy-4-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

37756-500MG 37756-1G 37756-5G 37756-25G
77326-494EA 232.78 USD
77326-494 77350-170 77362-790 77385-616
(3-Methoxy-4-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(3-Methoxy-4-methylphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₂
MW: 248.321 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20539823
CAS Number: 1378319-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR