tert-Butyl 4-chloro-3-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37750-500MG 37750-1G 37750-5G
77326-492EA 345.05 USD
77326-492 77350-168 77374-374
tert-Butyl 4-chloro-3-isopropylbenzoate ≥97%
tert-Butyl 4-chloro-3-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₉ClO₂
MW: 254.752 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403740
CAS Number: 2635937-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR