2,3-Difluoro-5-methoxy-4-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37742-500MG 37742-1G 37742-5G
77338-402EA 486.16 USD
77338-402 77350-160 77374-366
2,3-Difluoro-5-methoxy-4-(methylthio)benzaldehyde ≥97%
2,3-Difluoro-5-methoxy-4-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂S
MW: 218.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403737
CAS Number: 2624417-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR