(2,3-Difluoro-4-iodo-6-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

37738-500MG 37738-1G 37738-5G
77339-000EA 543.84 USD
77339-000 77350-156 77374-362
(2,3-Difluoro-4-iodo-6-methoxyphenyl)(methyl)sulfane ≥97%
(2,3-Difluoro-4-iodo-6-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₇F₂IOS
Storage Temperature: Ambient
MDL Number: MFCD33403734
CAS Number: 2624417-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR