(2,3-Difluoro-6-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

37737-500MG 37737-1G 37737-5G
77326-488EA 324.45 USD
77326-488 77350-154 77374-360
(2,3-Difluoro-6-methoxyphenyl)(methyl)sulfane ≥97%
(2,3-Difluoro-6-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈F₂OS
Storage Temperature: Ambient
MDL Number: MFCD33403733
CAS Number: 2386417-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR