Ethyl 2,3-difluoro-5-methoxy-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37733-500MG 37733-1G 37733-5G
77337-846EA 449.08 USD
77337-846 77350-150 77374-356
Ethyl 2,3-difluoro-5-methoxy-4-methylbenzoate ≥97%
Ethyl 2,3-difluoro-5-methoxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₃
MW: 230.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902614
CAS Number: 2586125-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR