2-Fluoro-4-(methoxymethyl)-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

37705-500MG 37705-1G 37705-5G 37705-25G
77326-480EA 92.7 USD
77326-480 77342-462 77362-784 77374-352
2-Fluoro-4-(methoxymethyl)-1-methylbenzene ≥95%
2-Fluoro-4-(methoxymethyl)-1-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO
MW: 154.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632264
CAS Number: 2404734-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR