5-Bromo-1-chloro-2-iodo-3-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

37704-500MG 37704-1G 37704-5G 37704-25G
77326-478EA 281.19 USD
77326-478 77350-144 77362-782 77385-608
5-Bromo-1-chloro-2-iodo-3-methoxybenzene ≥95%
5-Bromo-1-chloro-2-iodo-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClIO
MW: 347.375 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31558269
CAS Number: 1615212-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR