(2,6-Dichloro-4-methylphenyl)(methyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
37698-500MG
37698-1G
37698-5G
37698-25G
77326-476EA
165.83
USD
77326-476
77346-162
77362-780
77385-606
(2,6-Dichloro-4-methylphenyl)(methyl)sulfane ≥95%
(2,6-Dichloro-4-methylphenyl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₈H₈Cl₂S Storage Temperature: Ambient |
MDL Number:
MFCD28142022 CAS Number: 1804896-96-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...