Ethyl 2-bromo-6-fluoro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

37693-500MG 37693-1G 37693-25G 37693-5G
77326-474EA 304.88 USD
77326-474 77350-142 77385-604 77374-350
Ethyl 2-bromo-6-fluoro-3-methoxybenzoate ≥97%
Ethyl 2-bromo-6-fluoro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD33403732
CAS Number: 2166829-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR