Methyl 3-bromo-2-(difluoromethoxy)-5-isopropylbenzoate ≥95%

Supplier: AOBChem USA

Warning

37688-500MG 37688-1G 37688-5G
77339-182EA 560.32 USD
77339-182 77350-138 77374-346
Methyl 3-bromo-2-(difluoromethoxy)-5-isopropylbenzoate ≥95%
Methyl 3-bromo-2-(difluoromethoxy)-5-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrF₂O₃
MW: 323.131 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632259
CAS Number: 2404733-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR