3-Bromo-2-(difluoromethoxy)-5-isopropylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

37685-500MG 37685-1G 37685-5G
77339-588EA 602.55 USD
77339-588 77350-136 77374-344
3-Bromo-2-(difluoromethoxy)-5-isopropylbenzaldehyde ≥95%
3-Bromo-2-(difluoromethoxy)-5-isopropylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrF₂O₂
MW: 293.105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632257
CAS Number: 2404734-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR