Methyl 4-butoxy-2,6-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37682-500MG 37682-1G 37682-5G 37682-25G
77326-470EA 202.91 USD
77326-470 77350-134 77362-778 77385-600
Methyl 4-butoxy-2,6-dimethylbenzoate ≥97%
Methyl 4-butoxy-2,6-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₃
Storage Temperature: Ambient
MDL Number: MFCD33403730
CAS Number: 2624417-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR