Methyl 2,6-dimethyl-4-propoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

37676-500MG 37676-1G 37676-5G 37676-25G
77326-468EA 161.71 USD
77326-468 77345-856 77362-776 77385-598
Methyl 2,6-dimethyl-4-propoxybenzoate ≥97%
Methyl 2,6-dimethyl-4-propoxybenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD31773023
CAS Number: 1369403-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR