Methyl 5-propylthiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

37650-500MG 37650-1G 37650-5G 37650-25G
77326-462EA 243.08 USD
77326-462 77350-132 77362-770 77385-594
Methyl 5-propylthiophene-2-carboxylate ≥95%
Methyl 5-propylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD33403727
CAS Number: 161618-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR