Methyl 4,5-dichloro-2-fluoro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37646-500MG 37646-1G 37646-5G 37646-25G
77326-458EA 181.28 USD
77326-458 77346-830 77362-766 77385-590
Methyl 4,5-dichloro-2-fluoro-3-methylbenzoate ≥97%
Methyl 4,5-dichloro-2-fluoro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₇Cl₂FO₂
MW: 237.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902609
CAS Number: 2586126-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR