4,5-Dichloro-2-fluoro-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37645-500MG 37645-1G 37645-5G 37645-25G
77326-456EA 122.57 USD
77326-456 77343-990 77362-764 77374-340
4,5-Dichloro-2-fluoro-3-methylbenzoic acid ≥97%
4,5-Dichloro-2-fluoro-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅Cl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD32707042
CAS Number: 2432848-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR