4,5-Dichloro-2-fluoro-3-methylphenol ≥97%

Supplier: AOBChem USA

Warning

37643-500MG 37643-1G 37643-25G 37643-5G
77326-452EA 284.28 USD
77326-452 77350-126 77385-586 77374-338
4,5-Dichloro-2-fluoro-3-methylphenol ≥97%
4,5-Dichloro-2-fluoro-3-methylphenol

Precaución: for Research Use Only

Formula: C₇H₅Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD32707040
CAS Number: 2432849-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR