4-Bromo-6-fluoro-3-iodo-1H-indazole ≥97%
Supplier: AOBChem USA
|
Warning
|
37641-500MG
37641-1G
37641-5G
37641-25G
77326-450EA
127.72
USD
77326-450
77344-206
77362-760
77374-336
4-Bromo-6-fluoro-3-iodo-1H-indazole ≥97%
4-Bromo-6-fluoro-3-iodo-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₃BrFIN₂ Storage Temperature: Ambient |
MDL Number:
MFCD07781831 CAS Number: 887567-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...