(4-Fluoro-2-methoxy-3-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

37625-500MG 37625-1G 37625-5G
77338-410EA 487.19 USD
77338-410 77350-122 77374-332
(4-Fluoro-2-methoxy-3-methylphenyl)methanol ≥97%
(4-Fluoro-2-methoxy-3-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28519154
CAS Number: 1780709-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR