tert-Butyl 4-chloro-3,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37610-500MG 37610-1G 37610-5G 37610-25G
77326-446EA 168.92 USD
77326-446 77346-252 77362-758 77385-582
tert-Butyl 4-chloro-3,5-dimethylbenzoate ≥97%
tert-Butyl 4-chloro-3,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₇ClO₂
MW: 240.726 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403725
CAS Number: 2635937-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR