4-Isopropoxy-3,5-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

37600-500MG 37600-1G 37600-5G 37600-25G
77326-444EA 187.46 USD
77326-444 77350-118 77362-756 77385-580
4-Isopropoxy-3,5-dimethylphenol ≥97%
4-Isopropoxy-3,5-dimethylphenol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂
MW: 180.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044525
CAS Number: 1881296-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR