2,5-Dimethyl-4-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

37599-500MG 37599-1G 37599-5G 37599-25G
77326-442EA 201.88 USD
77326-442 77350-116 77362-754 77385-578
2,5-Dimethyl-4-(methylthio)benzoic acid ≥97%
2,5-Dimethyl-4-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂S
MW: 196.266 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902607
CAS Number: 2586126-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR