2,5-Dimethyl-4-(methylthio)benzaldehyde ≥95%
Supplier: AOBChem USA
|
Warning
|
37598-500MG
37598-1G
37598-5G
37598-25G
77326-440EA
202.91
USD
77326-440
77350-114
77362-752
77385-576
2,5-Dimethyl-4-(methylthio)benzaldehyde ≥95%
2,5-Dimethyl-4-(methylthio)benzaldehyde
Precaución: for Research Use Only
Formula:
C₁₀H₁₂OS MW: 180.267 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32902606 CAS Number: 2586125-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...