2-Bromo-1,3-dichloro-5-isopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

37595-500MG 37595-1G
77340-502EA 1313.25 USD
77340-502 77350-112
2-Bromo-1,3-dichloro-5-isopropylbenzene ≥95%
2-Bromo-1,3-dichloro-5-isopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₉BrCl₂
MW: 267.978 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34563124
CAS Number: 2731010-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR