1,2-Dichloro-4-fluoro-3-(methoxymethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

37579-500MG 37579-1G 37579-5G 37579-25G
77326-438EA 168.92 USD
77326-438 77346-326 77362-750 77385-574
1,2-Dichloro-4-fluoro-3-(methoxymethyl)benzene ≥95%
1,2-Dichloro-4-fluoro-3-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD32632242
CAS Number: 2404734-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR