4-Chloro-2,3-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

37573-500MG 37573-1G 37573-5G
77338-924EA 531.48 USD
77338-924 77350-110 77374-328
4-Chloro-2,3-dimethylphenol ≥97%
4-Chloro-2,3-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉ClO
MW: 156.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045775
CAS Number: 1570-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR