(2-Chloro-3-fluoro-4-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

37572-500MG 37572-1G 37572-5G
77338-374EA 484.1 USD
77338-374 77350-108 77374-326
(2-Chloro-3-fluoro-4-methoxyphenyl)boronic acid ≥97%
(2-Chloro-3-fluoro-4-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BClFO₃
MW: 204.391 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414603
CAS Number: 2282635-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR