1,5-Dibromo-3-chloro-2-(difluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

37560-500MG 37560-1G 37560-5G
77326-434EA 341.96 USD
77326-434 77350-106 77374-324
1,5-Dibromo-3-chloro-2-(difluoromethoxy)benzene ≥95%
1,5-Dibromo-3-chloro-2-(difluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃Br₂ClF₂O
MW: 336.356 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632235
CAS Number: 2385779-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR