4,7-Dibromo-3-iodo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

37555-500MG 37555-1G 37555-5G
77337-544EA 419.21 USD
77337-544 77350-102 77374-322
4,7-Dibromo-3-iodo-1H-indazole ≥97%
4,7-Dibromo-3-iodo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₃Br₂IN₂
Storage Temperature: Ambient
MDL Number: MFCD29089673
CAS Number: 2383369-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR