2-Fluoro-3-iodo-6-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37551-500MG 37551-1G 37551-5G 37551-25G
77326-430EA 161.71 USD
77326-430 77345-852 77362-744 77385-568
2-Fluoro-3-iodo-6-methoxybenzaldehyde ≥97%
2-Fluoro-3-iodo-6-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
Storage Temperature: Ambient
MDL Number: MFCD29922096
CAS Number: 1956319-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR