2-Fluoro-3-formyl-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37550-500MG 37550-1G 37550-5G 37550-25G
77326-428EA 120.51 USD
77326-428 77343-686 77362-742 77374-320
2-Fluoro-3-formyl-4-methoxybenzoic acid ≥97%
2-Fluoro-3-formyl-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₄
MW: 198.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756050
CAS Number: 2386892-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR