(4-Fluoro-2,5-dimethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

37513-500MG 37513-1G 37513-5G
77339-026EA 545.9 USD
77339-026 77350-096 77374-310
(4-Fluoro-2,5-dimethylphenyl)methanol ≥97%
(4-Fluoro-2,5-dimethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD28551041
CAS Number: 1785361-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR