Ethyl 2,3-difluoro-4-isopropoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

37511-500MG 37511-1G 37511-5G 37511-25G
77326-412EA 181.28 USD
77326-412 77346-828 77362-728 77385-560
Ethyl 2,3-difluoro-4-isopropoxybenzoate ≥95%
Ethyl 2,3-difluoro-4-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₂O₃
MW: 244.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902604
CAS Number: 2504203-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR