1-Bromo-3-ethoxy-5-(methoxymethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

37497-500MG 37497-1G 37497-5G 37497-25G
77326-410EA 218.36 USD
77326-410 77350-094 77362-724 77385-558
1-Bromo-3-ethoxy-5-(methoxymethyl)benzene ≥95%
1-Bromo-3-ethoxy-5-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₂
MW: 245.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632227
CAS Number: 2404733-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR