(3-Bromo-5-ethoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

37496-500MG 37496-1G 37496-5G 37496-25G
77326-408EA 181.28 USD
77326-408 77346-826 77362-722 77385-556
(3-Bromo-5-ethoxyphenyl)methanol ≥95%
(3-Bromo-5-ethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
MW: 231.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31637307
CAS Number: 1646557-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR