4-Bromo-2-fluoro-3-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37488-500MG 37488-1G 37488-25G 37488-5G
77326-406EA 285.31 USD
77326-406 77350-092 77385-554 77374-304
4-Bromo-2-fluoro-3-hydroxybenzaldehyde ≥97%
4-Bromo-2-fluoro-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28132109
CAS Number: 1805027-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR