(2,3-Difluoro-4-isopropoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

37486-500MG 37486-1G 37486-5G 37486-25G
77326-404EA 172.01 USD
77326-404 77346-414 77362-720 77385-552
(2,3-Difluoro-4-isopropoxyphenyl)methanol ≥97%
(2,3-Difluoro-4-isopropoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂O₂
MW: 202.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529936
CAS Number: 2734772-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR