2,3-Difluoro-4-isopropoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

37485-500MG 37485-1G 37485-5G 37485-25G
77326-402EA 144.2 USD
77326-402 77345-058 77362-718 77385-550
2,3-Difluoro-4-isopropoxybenzaldehyde ≥95%
2,3-Difluoro-4-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632226
CAS Number: 2404734-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR