(2,3-Difluoro-4-isopropoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

37482-1G 37482-5G 37482-25G
77340-658EA 60.77 USD
77340-658 77362-716 77374-302
(2,3-Difluoro-4-isopropoxyphenyl)(methyl)sulfane ≥95%
(2,3-Difluoro-4-isopropoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂OS
MW: 218.263 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403723
CAS Number: 2624417-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR