1-Chloro-2,3-difluoro-4-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

37480-500MG 37480-1G 37480-5G 37480-25G
77326-400EA 218.36 USD
77326-400 77350-090 77362-714 77385-548
1-Chloro-2,3-difluoro-4-isopropoxybenzene ≥97%
1-Chloro-2,3-difluoro-4-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉ClF₂O
MW: 206.617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044121
CAS Number: 1881293-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR