2,3-Difluoro-1-iodo-4-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

37479-500MG 37479-1G 37479-5G 37479-25G
77326-398EA 130.81 USD
77326-398 77344-326 77362-712 77385-546
2,3-Difluoro-1-iodo-4-isopropoxybenzene ≥97%
2,3-Difluoro-1-iodo-4-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉F₂IO
MW: 298.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403721
CAS Number: 2635937-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR