2,3-Difluoro-4-(hexyloxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37477-1G 37477-5G 37477-25G
77340-996EA 66.95 USD
77340-996 77362-710 77374-300
2,3-Difluoro-4-(hexyloxy)benzaldehyde ≥97%
2,3-Difluoro-4-(hexyloxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD32707033
CAS Number: 1277641-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR