(2,3-Difluoro-4-(hexyloxy)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

37473-500MG 37473-1G 37473-5G 37473-25G
77326-396EA 120.51 USD
77326-396 77343-684 77362-708 77374-298
(2,3-Difluoro-4-(hexyloxy)phenyl)(methyl)sulfane ≥97%
(2,3-Difluoro-4-(hexyloxy)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₃H₁₈F₂OS
Storage Temperature: Ambient
MDL Number: MFCD32707029
CAS Number: 2432848-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR