1-Chloro-2,3-difluoro-4-(hexyloxy)benzene ≥97%

Supplier: AOBChem USA

Warning

37471-500MG 37471-1G 37471-5G 37471-25G
77326-392EA 232.78 USD
77326-392 77350-088 77362-704 77385-542
1-Chloro-2,3-difluoro-4-(hexyloxy)benzene ≥97%
1-Chloro-2,3-difluoro-4-(hexyloxy)benzene

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD29044200
CAS Number: 1881288-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR