4-Butoxy-2,3-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37456-1G 37456-5G 37456-25G
77340-994EA 66.95 USD
77340-994 77362-700 77374-292
4-Butoxy-2,3-difluorobenzaldehyde ≥97%
4-Butoxy-2,3-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₂
MW: 214.209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529935
CAS Number: 853584-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR